Assay Details
| Gene Symbol: LOC4335530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781114.2 | XP_015636600.1 | 7 - 8 | 680 | 109 | |
| GenBank mRNA | AK064621.1 | - | 7 - 8 | 605 | 109 | |
| KX434730.1 | - | 7 - 8 | 457 | 109 | ||
More Information
Set Membership: |
|

LOC4335530
Genomic Map