Assay Details
Gene Symbol: LOC4335600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780778.2 | XP_015636264.1 | 7 - 8 | 1372 | 87 | |
GenBank mRNA | AK101615.1 | - | 7 - 8 | 1228 | 87 |
Gene Symbol: LOC4350816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK288325.1 | - | 1520 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
