Assay Details
Gene Symbol: LOC4335624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780857.1 | XP_015636343.1 | 3 - 4 | 1547 | 90 | |
GenBank mRNA | AK101106.1 | - | 3 - 4 | 1552 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
