Assay Details
| Gene Symbol: LOC4335896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780377.1 | XP_015635863.1 | 3 - 4 | 531 | 113 | |
| GenBank mRNA | AB114855.1 | - | 3 - 4 | 495 | 113 | |
| AK063934.1 | - | 3 - 4 | 503 | 113 | ||
| AK103344.1 | - | 3 - 4 | 504 | 113 | ||
More Information
Set Membership: |
|

LOC4335896
Genomic Map