Assay Details
| Gene Symbol: LOC4335896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780377.1 | XP_015635863.1 | 6 - 7 | 788 | 83 | |
| GenBank mRNA | AB114855.1 | - | 6 - 7 | 752 | 83 | |
| AK063934.1 | - | 6 - 7 | 760 | 83 | ||
| AK103344.1 | - | 6 - 7 | 761 | 83 | ||
More Information
Set Membership: |
|

LOC4335896
Genomic Map