Assay Details
| Gene Symbol: LOC4336257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780844.1 | XP_015636330.1 | 7 - 8 | 473 | 78 | |
| XM_015780845.2 | XP_015636331.1 | 5 - 6 | 694 | 78 | ||
| XM_015780846.1 | XP_015636332.1 | 6 - 7 | 419 | 78 | ||
| XM_015780847.1 | XP_015636333.1 | 7 - 8 | 473 | 78 | ||
| GenBank mRNA | AK067292.1 | - | 5 - 6 | 675 | 78 | |
More Information
Set Membership: |
|

LOC4336257
Genomic Map