Assay Details
Gene Symbol: LOC4336362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780807.2 | XP_015636293.1 | 15 - 16 | 2454 | 109 | |
XM_015780808.2 | XP_015636294.1 | 13 - 14 | 1846 | 109 | ||
GenBank mRNA | AB353922.1 | - | 15 - 16 | 2459 | 109 | |
AK066586.1 | - | 3 - 4 | 636 | 109 | ||
EU009924.1 | - | 16 - 17 | 2427 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
