Assay Details
Gene Symbol: LOC107275790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780952.2 | XP_015636438.1 | 1 - 2 | 279 | 73 | |
XM_015780955.2 | XP_015636441.1 | 1 - 2 | 366 | 73 | ||
GenBank mRNA | AK059412.1 | - | 1 - 2 | 208 | 73 | |
AK103012.1 | - | 1 - 2 | 369 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
