Assay Details
Gene Symbol: LOC107275790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780952.2 | XP_015636438.1 | 3 - 4 | 1125 | 92 | |
XM_015780953.2 | XP_015636439.1 | 3 - 4 | 1119 | 92 | ||
XM_015780955.2 | XP_015636441.1 | 3 - 4 | 1212 | 92 | ||
GenBank mRNA | AK059412.1 | - | 3 - 4 | 1053 | 92 | |
AK103012.1 | - | 3 - 4 | 1215 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
