Assay Details
Gene Symbol: LOC107275790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780955.2 | XP_015636441.1 | 5 - 6 | 1492 | 126 | |
GenBank mRNA | AK103012.1 | - | 5 - 6 | 1495 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
