Assay Details
| Gene Symbol: LOC4336547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780081.2 | XP_015635567.1 | 6 - 7 | 1309 | 82 | |
| GenBank mRNA | AK068610.1 | - | 6 - 7 | 1269 | 82 | |
| AY224498.1 | - | 1 - 2 | 600 | 82 | ||
More Information
Set Membership: |
|

LOC4336547
Genomic Map