Assay Details
| Gene Symbol: LOC4336584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780265.2 | XP_015635751.1 | 3 - 4 | 473 | 94 | |
| XM_015780266.2 | XP_015635752.1 | 3 - 4 | 432 | 94 | ||
| GenBank mRNA | AK061922.1 | - | 2 - 3 | 251 | 94 | |
| AK101956.1 | - | 3 - 4 | 429 | 94 | ||
| EU325987.1 | - | 2 - 3 | 311 | 94 | ||
More Information
Set Membership: |
|

LOC4336584
Genomic Map