Assay Details
Gene Symbol: LOC4336589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780898.2 | XP_015636384.1 | 15 - 16 | 2456 | 126 | |
GenBank mRNA | AK067475.1 | - | 15 - 16 | 1969 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
