Assay Details
Gene Symbol: LOC4336589 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780898.2 | XP_015636384.1 | 4 - 5 | 1047 | 78 | |
GenBank mRNA | AK067475.1 | - | 4 - 5 | 539 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
