Assay Details
| Gene Symbol: LOC4336982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780230.2 | XP_015635716.1 | 4 - 5 | 438 | 142 | |
| XM_015780231.2 | XP_015635717.1 | 4 - 5 | 528 | 142 | ||
| XM_015780232.2 | XP_015635718.1 | 4 - 5 | 557 | 142 | ||
| GenBank mRNA | AK100259.1 | - | 4 - 5 | 558 | 142 | |
More Information
Set Membership: |
|

LOC4336982
Genomic Map