Assay Details
Gene Symbol: LOC4337656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785337.2 | XP_015640823.1 | 1 - 2 | 352 | 114 | |
XM_015785338.2 | XP_015640824.1 | 1 - 2 | 357 | 114 | ||
GenBank mRNA | AK101222.1 | - | 1 - 2 | 230 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
