Assay Details
Gene Symbol: LOC4337656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785337.2 | XP_015640823.1 | 2 - 3 | 554 | 103 | |
XM_015785338.2 | XP_015640824.1 | 2 - 3 | 559 | 103 | ||
XM_015785340.2 | XP_015640826.1 | 4 - 5 | 910 | 103 | ||
XM_015785346.2 | XP_015640832.1 | 5 - 6 | 911 | 103 | ||
GenBank mRNA | AK101222.1 | - | 2 - 3 | 432 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
