Assay Details
Gene Symbol: LOC4337656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785337.2 | XP_015640823.1 | 3 - 4 | 838 | 99 | |
XM_015785338.2 | XP_015640824.1 | 3 - 4 | 843 | 99 | ||
XM_015785340.2 | XP_015640826.1 | 5 - 6 | 1194 | 99 | ||
XM_015785346.2 | XP_015640832.1 | 6 - 7 | 1195 | 99 | ||
GenBank mRNA | AK101222.1 | - | 3 - 4 | 716 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
