Assay Details
Gene Symbol: LOC4337656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785337.2 | XP_015640823.1 | 4 - 5 | 867 | 78 | |
XM_015785338.2 | XP_015640824.1 | 4 - 5 | 872 | 78 | ||
XM_015785340.2 | XP_015640826.1 | 6 - 7 | 1223 | 78 | ||
XM_015785346.2 | XP_015640832.1 | 7 - 8 | 1224 | 78 | ||
GenBank mRNA | AK101222.1 | - | 4 - 5 | 745 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
