Assay Details
Gene Symbol: LOC4337656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785338.2 | XP_015640824.1 | 7 - 8 | 1673 | 74 | |
GenBank mRNA | AK101222.1 | - | 7 - 8 | 1546 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
