Assay Details
| Gene Symbol: LOC4338726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784214.2 | XP_015639700.1 | 2 - 3 | 365 | 70 | |
| GenBank mRNA | AK072739.1 | - | 2 - 3 | 358 | 70 | |
| AK103913.1 | - | 2 - 3 | 357 | 70 | ||
| BT129077.1 | - | 2 - 3 | 351 | 70 | ||
More Information
Set Membership: |
|

LOC4338726
Genomic Map