Assay Details
| Gene Symbol: LOC4337299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780938.2 | XP_015636424.1 | 5 - 6 | 542 | 79 | |
| XM_015780939.2 | XP_015636425.1 | 5 - 6 | 476 | 79 | ||
| GenBank mRNA | AK061921.1 | - | 5 - 6 | 472 | 79 | |
| AK065171.1 | - | 5 - 6 | 479 | 79 | ||
| AK099504.1 | - | 5 - 6 | 480 | 79 | ||
More Information
Set Membership: |
|

LOC4337299
Genomic Map