Assay Details
| Gene Symbol: LOC4337415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778380.2 | XP_015633866.1 | 1 - 2 | 123 | 64 | |
| GenBank mRNA | AK065236.1 | - | 1 - 2 | 130 | 64 | |
| AK103940.1 | - | 1 - 2 | 129 | 64 | ||
| AK103943.1 | - | 1 - 2 | 129 | 64 | ||
| AK104855.1 | - | 1 - 2 | 129 | 64 | ||
More Information
Set Membership: |
|

LOC4337415
Genomic Map