Assay Details
Gene Symbol: LOC4337452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781182.1 | XP_015636668.1 | 5 - 6 | 782 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
