Assay Details
| Gene Symbol: LOC4337825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783895.1 | XP_015639381.1 | 5 - 6 | 1139 | 96 | |
| GenBank mRNA | AK100732.1 | - | 5 - 6 | 1146 | 96 | |
More Information
Set Membership: |
|

LOC4337825
Genomic Map