Assay Details
| Gene Symbol: LOC4338067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782574.2 | XP_015638060.1 | 6 - 7 | 813 | 84 | |
| GenBank mRNA | AB232157.1 | - | 5 - 6 | 658 | 84 | |
| AK111723.1 | - | 5 - 6 | 658 | 84 | ||
More Information
Set Membership: |
|

LOC4338067
Genomic Map