Assay Details
| Gene Symbol: LOC4339007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782700.2 | XP_015638186.1 | 11 - 12 | 2033 | 96 | |
| XM_015782701.2 | XP_015638187.1 | 11 - 12 | 1756 | 96 | ||
| GenBank mRNA | AK073413.1 | - | 11 - 12 | 1763 | 96 | |
| AK104040.1 | - | 9 - 10 | 1242 | 96 | ||
More Information
Set Membership: |
|

LOC4339007
Genomic Map