Assay Details
| Gene Symbol: LOC4339007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782701.2 | XP_015638187.1 | 7 - 8 | 1387 | 83 | |
| GenBank mRNA | AK073413.1 | - | 7 - 8 | 1394 | 83 | |
| AK104040.1 | - | 5 - 6 | 873 | 83 | ||
More Information
Set Membership: |
|

LOC4339007
Genomic Map