Assay Details
| Gene Symbol: LOC4339007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782700.2 | XP_015638186.1 | 8 - 9 | 1892 | 137 | |
| XM_015782701.2 | XP_015638187.1 | 8 - 9 | 1615 | 137 | ||
| GenBank mRNA | AK073413.1 | - | 8 - 9 | 1622 | 137 | |
| AK104040.1 | - | 6 - 7 | 1101 | 137 | ||
More Information
Set Membership: |
|

LOC4339007
Genomic Map