Assay Details
| Gene Symbol: LOC4339189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783826.2 | XP_015639312.1 | 1 - 2 | 345 | 80 | |
| GenBank mRNA | AK068320.1 | - | 1 - 2 | 152 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015783826.2 transcripts.
Set Membership: |
|

LOC4339189
Genomic Map