Assay Details
| Gene Symbol: LOC4339273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784725.2 | XP_015640211.1 | 1 - 2 | 282 | 105 | |
| XM_015784726.2 | XP_015640212.1 | 1 - 2 | 283 | 105 | ||
| GenBank mRNA | AK108053.1 | - | 1 - 2 | 276 | 105 | |
More Information
Set Membership: |
|

LOC4339273
Genomic Map