Assay Details
| Gene Symbol: LOC4339273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784725.2 | XP_015640211.1 | 3 - 4 | 527 | 92 | |
| GenBank mRNA | AK108053.1 | - | 3 - 4 | 521 | 92 | |
More Information
Set Membership: |
|

LOC4339273
Genomic Map