Assay Details
Gene Symbol: LOC4340043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788881.2 | XP_015644367.1 | 25 - 26 | 3490 | 62 | |
XM_015788882.2 | XP_015644368.1 | 25 - 26 | 3487 | 62 | ||
XM_015788883.2 | XP_015644369.1 | 25 - 26 | 3484 | 62 | ||
XM_015788884.2 | XP_015644370.1 | 23 - 24 | 3124 | 62 | ||
XM_015788885.2 | XP_015644371.1 | 25 - 26 | 3375 | 62 | ||
XM_015788886.2 | XP_015644372.1 | 26 - 27 | 3287 | 62 | ||
GenBank mRNA | AK065081.1 | - | 21 - 22 | 4990 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
