Assay Details
Gene Symbol: LOC4340043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788881.2 | XP_015644367.1 | 2 - 3 | 388 | 70 | |
XM_015788882.2 | XP_015644368.1 | 2 - 3 | 388 | 70 | ||
XM_015788883.2 | XP_015644369.1 | 2 - 3 | 388 | 70 | ||
XM_015788884.2 | XP_015644370.1 | 2 - 3 | 388 | 70 | ||
GenBank mRNA | AK065081.1 | - | 2 - 3 | 367 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
