Assay Details
Gene Symbol: LOC4340043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788881.2 | XP_015644367.1 | 3 - 4 | 739 | 67 | |
XM_015788882.2 | XP_015644368.1 | 3 - 4 | 739 | 67 | ||
XM_015788883.2 | XP_015644369.1 | 3 - 4 | 739 | 67 | ||
XM_015788884.2 | XP_015644370.1 | 3 - 4 | 739 | 67 | ||
XM_015788885.2 | XP_015644371.1 | 3 - 4 | 624 | 67 | ||
XM_015788886.2 | XP_015644372.1 | 4 - 5 | 536 | 67 | ||
GenBank mRNA | AK065081.1 | - | 3 - 4 | 718 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788886.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
