Assay Details
Gene Symbol: LOC4340043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788881.2 | XP_015644367.1 | 5 - 6 | 984 | 116 | |
XM_015788882.2 | XP_015644368.1 | 5 - 6 | 984 | 116 | ||
XM_015788883.2 | XP_015644369.1 | 5 - 6 | 984 | 116 | ||
XM_015788884.2 | XP_015644370.1 | 5 - 6 | 984 | 116 | ||
XM_015788885.2 | XP_015644371.1 | 5 - 6 | 869 | 116 | ||
XM_015788886.2 | XP_015644372.1 | 6 - 7 | 781 | 116 | ||
GenBank mRNA | AK065081.1 | - | 5 - 6 | 963 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
