Assay Details
Gene Symbol: LOC4341063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785665.2 | XP_015641151.1 | 1 - 2 | 363 | 89 | |
GenBank mRNA | AK062871.1 | - | 1 - 2 | 352 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
