Assay Details
Gene Symbol: LOC4342038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789007.2 | XP_015644493.1 | 1 - 2 | 307 | 85 | |
GenBank mRNA | AK064384.1 | - | 1 - 2 | 300 | 85 | |
AK102831.1 | - | 1 - 2 | 273 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
