Assay Details
Gene Symbol: LOC4342038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789007.2 | XP_015644493.1 | 4 - 5 | 860 | 100 | |
GenBank mRNA | AK064384.1 | - | 4 - 5 | 853 | 100 | |
AK102831.1 | - | 4 - 5 | 826 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
