Assay Details
Gene Symbol: LOC4342437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791637.2 | XP_015647123.1 | 8 - 9 | 829 | 102 | |
XM_015791638.2 | XP_015647124.1 | 9 - 10 | 706 | 102 | ||
GenBank mRNA | AK103532.1 | - | 9 - 10 | 688 | 102 | |
AK103591.1 | - | 9 - 10 | 688 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
