Assay Details
| Gene Symbol: LOC4343080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789420.2 | XP_015644906.1 | 11 - 12 | 1362 | 108 | |
| GenBank mRNA | AK060367.1 | - | 3 - 4 | 236 | 108 | |
| AK103253.1 | - | 11 - 12 | 1342 | 108 | ||
More Information
Set Membership: |
|

LOC4343080
Genomic Map