Assay Details
| Gene Symbol: LOC4343080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789420.2 | XP_015644906.1 | 5 - 6 | 753 | 119 | |
| GenBank mRNA | AK103253.1 | - | 5 - 6 | 733 | 119 | |
More Information
Set Membership: |
|

LOC4343080
Genomic Map