Assay Details
| Gene Symbol: LOC4343299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791108.2 | XP_015646594.1 | 6 - 7 | 3933 | 122 | |
| XM_015791109.2 | XP_015646595.1 | 6 - 7 | 4139 | 122 | ||
| XM_015791110.2 | XP_015646596.1 | 6 - 7 | 3876 | 122 | ||
| XM_015791111.2 | XP_015646597.1 | 6 - 7 | 3936 | 122 | ||
| XM_015791112.2 | XP_015646598.1 | 6 - 7 | 3816 | 122 | ||
| GenBank mRNA | AK100332.1 | - | 1 - 2 | 1634 | 122 | |
More Information
Set Membership: |
|

LOC4343299
Genomic Map