Assay Details
Gene Symbol: LOC4343299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791108.2 | XP_015646594.1 | 9 - 10 | 4280 | 69 | |
XM_015791109.2 | XP_015646595.1 | 9 - 10 | 4486 | 69 | ||
XM_015791110.2 | XP_015646596.1 | 9 - 10 | 4223 | 69 | ||
XM_015791111.2 | XP_015646597.1 | 9 - 10 | 4283 | 69 | ||
XM_015791112.2 | XP_015646598.1 | 9 - 10 | 4163 | 69 | ||
GenBank mRNA | AK100332.1 | - | 4 - 5 | 1995 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
