Assay Details
| Gene Symbol: LOC4343406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789221.2 | XP_015644707.1 | 2 - 3 | 4449 | 148 | |
| GenBank mRNA | AK106593.1 | - | 2 - 3 | 1453 | 148 | |
More Information
Set Membership: |
|

LOC4343406
Genomic Map