Assay Details
Gene Symbol: LOC4344339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792307.2 | XP_015647793.1 | 1 - 2 | 291 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792307.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
