Assay Details
Gene Symbol: LOC4344339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792305.2 | XP_015647791.1 | 2 - 3 | 406 | 81 | |
GenBank mRNA | AK067620.1 | - | 2 - 3 | 460 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
