Assay Details
Gene Symbol: LOC4344339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792305.2 | XP_015647791.1 | 3 - 4 | 512 | 91 | |
XM_015792306.2 | XP_015647792.1 | 2 - 3 | 292 | 91 | ||
XM_015792307.2 | XP_015647793.1 | 3 - 4 | 551 | 91 | ||
GenBank mRNA | AK067620.1 | - | 3 - 4 | 566 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
