Assay Details
Gene Symbol: LOC4345241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795318.2 | XP_015650804.1 | 1 - 2 | 396 | 148 | |
GenBank mRNA | AK121969.1 | - | 1 - 2 | 245 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() |
