Assay Details
Gene Symbol: LOC4345241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795318.2 | XP_015650804.1 | 5 - 6 | 649 | 139 | |
XM_015795319.2 | XP_015650805.1 | 5 - 6 | 866 | 139 | ||
GenBank mRNA | AK121969.1 | - | 5 - 6 | 498 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
